Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-3CL-PRO Rabbit Polyclonal Antibody

Supplier: Rockland Immunochemicals
200-401-A51 200-401-A51
RL200-401-A51EA 554 USD InStock
RL200-401-A51 CA200-401-A51
Anti-3CL-PRO Rabbit Polyclonal Antibody
Antibodies
This Protein A purified antibody has been tested for use in ELISA and by western blot.  Specific conditions for reactivity should be optimized by the end user. Expect a band approximately 34 kDa in size corresponding to SARS 3CL Protease by western blottiing.

Recommended Dilutions: ELISA: 1:10000 - 1:50000; Western Blot: 1:2000 - 1:10000
Restore with deionized water (or equivalent); 100 µl

Type: Primary
Antigen: 3CL-PRO
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: Full length
Host: Rabbit
Isotype: IgG
Reactivity: SARS Virus

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty