Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-IL13 Rabbit Polyclonal Antibody

Supplier: Rockland Immunochemicals
200-401-B55 200-401-B55S
RL200-401-B55SEA 148 USD InStock
RL200-401-B55S RL200-401-B55
Anti-IL13 Rabbit Polyclonal Antibody
Antibodies
This Protein-A purified antibody has been tested for use in ELISA and western blotting. Specific conditions for reactivity should be optimized by the end user. Expect a band approximately 13.2 kDa in size corresponding to swine IL-13 Protein by western blotting.

Recommended Dilutions: ELISA: 1:10000; Western Blot: 2 µg/ml;

Type: Primary
Antigen: IL13
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: Full length
Host: Rabbit
Isotype: IgG
Reactivity: Pig, Sheep, Bovine, Pig

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty