Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-ErpD Rabbit Polyclonal Antibody

Supplier: Rockland Immunochemicals
200-401-C09 200-401-C09S
RL200-401-C09SEA 148 USD InStock
RL200-401-C09S RL200-401-C09
Anti-ErpD Rabbit Polyclonal Antibody
Antibodies
Anti-ErpD antibody has been tested in Western Blot. Specific conditions for reactivity should be optimized by the end user. Expect bands at ~37.5 kDa and ~65 kDa in size corresponding to ErpD/Arp37 by Western blotting in the appropriate cell lysate or extract.

Recommended Dilutions: ELISA: >1:5000; Western Blot: 1:1000;

Type: Primary
Antigen: ErpD
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Borrelia burgdorferi

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty