Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-OSPA Rabbit Polyclonal Antibody

Supplier: Rockland Immunochemicals
200-401-C13S 200-401-C13
RL200-401-C13SEA 148 USD InStock
RL200-401-C13S RL200-401-C13
Anti-OSPA Rabbit Polyclonal Antibody
Antibodies
This Protein-A purified antibody has been tested for use in Western blotting. Specific conditions for reactivity should be optimized by the user. Expect a band approximately 28.1 kDa in size corresponding to Borrelia burgdorferi OspA Protein by western blotting.

Recommended Dilutions: ELISA: >1:5000; Western Blot: 1:1000;

Type: Primary
Antigen: OSPA
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Borrelia burgdorferi

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty