Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-BBA060 Rabbit Polyclonal Antibody

Supplier: Rockland Immunochemicals
200-401-C30 200-401-C30S
RL200-401-C30EA 518 USD InStock
RL200-401-C30 RL200-401-C30S
Anti-BBA060 Rabbit Polyclonal Antibody
Antibodies
Anti-Surface LipoProtein p27 antibody has been tested in Western Blot. Specific conditions for reactivity should be optimized by the end user. Expect a band at ~30.9 kDa in size corresponding to p27 by Western blotting in the appropriate cell lysate or extract.

Recommended Dilutions: ELISA: >1:5000; Western Blot: 1:1000;

Type: Primary
Antigen: BBA060
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Borrelia burgdorferi

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty