Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-VLSE Rabbit Polyclonal Antibody

Supplier: Rockland Immunochemicals
200-401-C33 200-401-C33S
RL200-401-C33EA 504 USD InStock
RL200-401-C33 RL200-401-C33S
Anti-VLSE Rabbit Polyclonal Antibody
Antibodies
Anti-VlsE antibody has been tested in ELISA and Western Blot. Specific conditions for reactivity should be optimized by the end user. Expect a band at ~36.3 kDa in size corresponding to VlsE by Western blotting in the appropriate cell lysate or extract.

Recommended Dilutions: ELISA: 1:250; Western Blot: 1:1000;

Type: Primary
Antigen: VLSE
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Borrelia burgdorferi

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty