Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-OLIG2 Rabbit Polyclonal Antibody

Supplier: Rockland Immunochemicals
200-401-E03
RL200-401-E03EA 518 USD InStock
RL200-401-E03
Anti-OLIG2 Rabbit Polyclonal Antibody
Antibodies
Anti-Olig2 Antibody is suitable for use in Western Blotting, Immunoprecipitation, Immunochemistryand ICC. Expect a band of approximately 32 kDa in size corresponding to Olig2 Proteins in Western blots in the appropriate cell lysate or extracts.

  • Recommended Dilutions: Western Blot: 1:3000; Immunochemistry: 1:1000; Immunoprecipitation: 1:500-1000

Type: Primary
Antigen: OLIG2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse, Human, Rat

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty