Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-APP Rabbit Polyclonal Antibody

Supplier: Rockland Immunochemicals
200-401-E88
RL200-401-E88EA 501 USD InStock
RL200-401-E88
Anti-APP Rabbit Polyclonal Antibody
Antibodies
Amyloid Oligomers (A11) Antibody

  • Recommended Dilutions: ELISA: 0.1-10 µg/ml; Western Blot: 1:1000; Immunochemistry: 1:1000-10000; Immunoprecipitation: 1:1000

Type: Primary
Antigen: APP
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse, Rat, Human

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty