Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-AGAP4 Rabbit Polyclonal Antibody

Supplier: Rockland Immunochemicals
200-401-P07S 200-401-P07S
RL200-401-P07SEA 148 USD InStock
CA200-401-P07S RL200-401-P07S
Anti-AGAP4 Rabbit Polyclonal Antibody
Antibodies
Anti-AGAP4 antibody has been tested for use in western blot. Specific conditions for reactivity should be optimized by the end user. Expect a band approximately 73kDa in size corresponding to AGAP4 by western blotting in the appropriate cell lysate.

Recommended Dilutions: ELISA: 1:10000-1:20000; Western Blot: 1:1000

Type: Primary
Antigen: AGAP4
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty