Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-ORM1 Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))

Supplier: Rockland Immunochemicals
200-403-046S 200-403-046S
RL200-403-046SEA 148 USD InStock
CA10036-984 RL200-403-046S
Anti-ORM1 Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))
Antibodies
Anti-Alpha-1-Acid GlycoProtein Antibody is suitable for ELISA, Western Blot and immunochemistry. Specific conditions for reactivity should be optimized by the end user.

Recommended Dilutions: ELISA: 1:15000; Western Blot: 1:1000 - 1:5000; Immunochemistry: 1:500 - 1:2500;
Do not add Sodium Azide!

Type: Primary
Antigen: ORM1
Clonality: Polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty