Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-CA2 Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))

Supplier: Rockland Immunochemicals
200-403-136S 200-403-136S
RL200-403-136SEA 148 USD InStock
RL200-403-136S CA10037-004
Anti-CA2 Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))
Antibodies
Anti-Carbonic anhydrase II has been assayed against 1.0 µg of Carbonic Anhydrase II (Human Erythrocytes) in a standard capture ELISA using ABTS (2,2’-azino-bis-[3-ethylbenthiazoline-6-sulfonic acid])

Recommended Dilutions: ELISA: 1:500 - 1:2500; Western Blot: 1:250 - 1:1000; Immunochemistry: 1:100 - 1:500;
Do not add Sodium Azide!

Type: Primary
Antigen: CA2
Clonality: Polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty