Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-Streptavidin Rabbit Polyclonal Antibody (Biotin)

Supplier: Rockland Immunochemicals
200-406-095S 200-406-095S
RL200-406-095SEA 148 USD InStock
RL200-406-095S CA200-406-095S
Anti-Streptavidin Rabbit Polyclonal Antibody (Biotin)
Antibodies
Biotin Conjugated Anti-Streptavidin Antibody has been assayed by ELISA for the detection of streptavidin in a standard ELISA using Peroxidase as a reporter.

Recommended Dilutions: ELISA: 1:2000-1:20000;

Type: Tag
Antigen: Streptavidin
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype:
Reactivity:

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty