Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-Protein G Rabbit Polyclonal Antibody (Biotin)

Supplier: Rockland Immunochemicals
200-406-138S 200-406-138S
RL200-406-138SEA 148 USD InStock
RL200-406-138S CA200-406-138S
Anti-Protein G Rabbit Polyclonal Antibody (Biotin)
Antibodies
Anti-Protein G Biotin Conjugated Antibody has been assayed against 1.0 µg of Protein G in a standard capture ELISA using Peroxidase Conjugated Streptavidin and ABTS (2,2’-azino-bis-[3-ethylbenthiazoline-6-sulfonic acid])

Recommended Dilutions: ELISA: 1:90,000-1:350000

Type: Tag
Antigen: Protein G
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype:
Reactivity:

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty