Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-Protein G Rabbit Polyclonal Antibody (Texas Red®)

Supplier: Rockland Immunochemicals
200-4091380100 200-409-138-0100
RL200-40913801EA 339 USD InStock
CA200409138010 RL200-40913801
Anti-Protein G Rabbit Polyclonal Antibody (Texas Red®)
Antibodies
Type: Tag
Antigen: Protein G
Clonality: Polyclonal
Clone:
Conjugation: Texas Red®
Epitope:
Host: Rabbit
Isotype:
Reactivity:

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty