Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-HEX Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))

Supplier: Rockland Immunochemicals
200-4359S 200-4359S
RL200-4359SEA 148 USD InStock
CA10037-082 RL200-4359S
Anti-HEX Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))
Antibodies
Anti-HEXOKINASE (RABBIT) Antibody can be used in applications such as immunochemistry , Western blot (WB) Immunofluorescence (IF) ELISA. Specific conditions should be optimized by researcher.

Recommended Dilutions: ELISA: 1:1000 - 1:2500; Western Blot: 1:500 - 1:1000;
Do not add Sodium Azide!

Type: Primary
Antigen: HEX
Clonality: Polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Rabbit
Isotype:
Reactivity: Yeast

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty