Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-Avidin Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))

Supplier: Rockland Immunochemicals
200-4397-0100 200-4397-0100
RL200-4397-01EA 467 USD InStock
CA20043970100 RL200-4397-01
Anti-Avidin Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))
Antibodies
Anti-Avidin Antibody Peroxidase Conjugated has been assayed against 1.0 ug of Avidin [Hen Egg White ] in a standard capture ELISA using ABTS (2,2’-azino-bis-[3-ethylbenthiazoline-6-sulfonic acid]) code # ABTS-100 as a substrate for 30 minutes at room temp

  • Recommended Dilutions: ELISA: 1:10000 - 1:50000; Immunofluorescence: 1:500 - 1:2000; Western Blot: 1:2000 - 1:5000

Type: Tag
Antigen: Avidin
Clonality: Polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Rabbit
Isotype:
Reactivity: 0

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty