Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-Avidin Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))

Supplier: Rockland Immunochemicals
200-4397S 200-4397S
RL200-4397SEA 148 USD InStock
CA200-4397S RL200-4397S
Anti-Avidin Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))
Antibodies
Anti-Avidin Antibody Peroxidase Conjugated has been assayed against 1.0 µg of Avidin (Hen Egg White ) in a standard capture ELISA using ABTS (2,2’-azino-bis-[3-ethylbenthiazoline-6-sulfonic acid])

  • Recommended Dilutions: ELISA: 1:10000 - 1:50000; Western Blot: 1:2000 - 1:5000; Immunochemistry: 1:500 - 1:2000
  • Do not add Sodium Azide!

Type: Tag
Antigen: Avidin
Clonality: Polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Rabbit
Isotype:
Reactivity: 0

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty