Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-AHSA1 Rat Monoclonal Antibody [clone: 25F2.D9]

Supplier: Rockland Immunochemicals
200-501-974
RL200-501-974EA 528 USD InStock
RL200-501-974
Anti-AHSA1 Rat Monoclonal Antibody [clone: 25F2.D9]
Antibodies
This Protein-G purified antibody has been tested for use in ELISA, immunochemistry and western blotting.  Specific conditions for reactivity should be optimized by the end user.  Expect a band approximately 38-40 kDa in size.

Recommended Dilutions: ELISA: 1:20000; Western Blot: 1:1000; Immunochemistry: 5-10 µg/ml;

Type: Primary
Antigen: AHSA1
Clonality: Monoclonal
Clone: 25F2.D9
Conjugation: Unconjugated
Epitope: Full length
Host: Rat
Isotype: IgG2a kappa
Reactivity: Mouse, Human, Chimpanzee, Mouse

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty