Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-GDF15 Rat Monoclonal Antibody [clone: 6D12.H10.E4]

Supplier: Rockland Immunochemicals
200-501-B07S 200-501-B07
RL200-501-B07EA 409 USD InStock
RL200-501-B07 RL200-501-B07S
Anti-GDF15 Rat Monoclonal Antibody [clone: 6D12.H10.E4]
Antibodies
This Protein G purified antibody is suitable for ELISA and western blotting of human NAG-1 Protein.  For detection of NAG-1 in human serum, a sandwich ELISA is suggested using this antibody in combination with anti-NAG-1/GDF15 (N-terminal), H variant or D.

Recommended Dilutions: ELISA: 1:2000; Western Blot: 1:1000;

Type: Primary
Antigen: GDF15
Clonality: Monoclonal
Clone: 6D12.H10.E4
Conjugation: Unconjugated
Epitope: C-terminal
Host: Rat
Isotype: IgG2a lambda
Reactivity: Human, Mouse

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty