Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-HSP90AB1 Rat Monoclonal Antibody [clone: 9G10]

Supplier: Rockland Immunochemicals
200-501-F39
RL200-501-F39EA 532 USD InStock
RL200-501-F39
Anti-HSP90AB1 Rat Monoclonal Antibody [clone: 9G10]
Antibodies
Anti-Grp94 Antibody is suitable for WB, IP, and Flow cytometry. Expect a band approximately 98kDa Protein corresponding to the molecular mass of Grp94 on SDS PAGE immunoblots. This monoclonal antibody reacts with purified Grp94.

Recommended Dilutions: Western Blot: 0.5µg/ml;

Type: Primary
Antigen: HSP90AB1
Clonality: Monoclonal
Clone: 9G10
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity: Human, Mouse, Rat, Bovine, Dog, Chicken, Guinea Pig, Hamster, Horse, Monkey, Pig, Rabbit, Sheep, Xenopus

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty