Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-HSF1 Rat Monoclonal Antibody [clone: 10H8]

Supplier: Rockland Immunochemicals
200-501-F50
RL200-501-F50EA 532 USD InStock
RL200-501-F50
Anti-HSF1 Rat Monoclonal Antibody [clone: 10H8]
Antibodies
Anti-HSF1 Antibody is suitable for WB, IP, Immunofluorescence microscopy, ELISA, and Gel mobility shift assay. Expect a band approximately ~85kDa Protein in unstressed cell lysates, and an ~95kDa Protein in heat shocked cell lysates.

Recommended Dilutions: ELISA: 1:200; Western Blot: 1 µg/ml;

Type: Primary
Antigen: HSF1
Clonality: Monoclonal
Clone: 10H8
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG1
Reactivity: Human, Mouse, Rat, Bovine, Guinea Pig, Hamster, Monkey, Rabbit

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty