Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-LY6C1 Rat Monoclonal Antibody (FITC (Fluorescein Isothiocyanate)) [clone: RB6-8C5]

Supplier: Rockland Immunochemicals
200-502-L54 200-502-L54
RL200-502-L54EA 550 USD InStock
CA200-502-L54 RL200-502-L54
Anti-LY6C1 Rat Monoclonal Antibody (FITC (Fluorescein Isothiocyanate)) [clone: RB6-8C5]
Antibodies
Anti-Ly-6G/Ly-6C (Gr-1) is useful for immunochemistry, Immunoprecipitation, Western Blot, and Flow Cytometry using mouse spleen cells, or an appropriate cell type (where indicated).

Recommended Dilutions: Flow Cytometry: 10 µl/10^6 cells (0.1 µg);
Do not freeze!

Type: Primary
Antigen: LY6C1
Clonality: Monoclonal
Clone: RB6-8C5
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Rat
Isotype: IgG2b kappa
Reactivity: Mouse

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty