Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-CD19 Rat Monoclonal Antibody (FITC (Fluorescein Isothiocyanate)) [clone: 1D3]

Supplier: Rockland Immunochemicals
200-502-M72 200-502-M72
RL200-502-M72EA 524 USD InStock
RL200-502-M72 CA10037-148
Anti-CD19 Rat Monoclonal Antibody (FITC (Fluorescein Isothiocyanate)) [clone: 1D3]
Antibodies
Anti-CD19 is useful for Flow Cytometry (Cell Surface). Researchers should determine optimal titers for applications that are not stated.

Recommended Dilutions: Flow Cytometry: 0.25-1µg/10^6 cells
Do not freeze!

Type: Primary
Antigen: CD19
Clonality: Monoclonal
Clone: 1D3
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Rat
Isotype:
Reactivity: Mouse

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty