Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-CD4 Rat Monoclonal Antibody (RPE (R-Phycoerythrin)) [clone: GK1.5]

Supplier: Rockland Immunochemicals
200-508-M70 200-508-M70
RL200-508-M70EA 285 USD InStock
CA10037-152 RL200-508-M70
Anti-CD4 Rat Monoclonal Antibody (RPE (R-Phycoerythrin)) [clone: GK1.5]
Antibodies
Anti-CD4 is useful for Immunofluorescence and Flow Cytometry (Cell Surface). Researchers should determine optimal titers for applications that are not stated.

Recommended Dilutions: Flow Cytometry: 0.1-1 µg/10^6 cells
Do not freeze! Light sensitive!

Type: Primary
Antigen: CD4
Clonality: Monoclonal
Clone: GK1.5
Conjugation: RPE (R-Phycoerythrin)
Epitope:
Host: Rat
Isotype: IgG2b kappa
Reactivity:

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty