Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-CD19 Rat Monoclonal Antibody (RPE (R-Phycoerythrin)) [clone: 1D3]

Supplier: Rockland Immunochemicals
200-508-M72 200-508-M72
RL200-508-M72EA 313 USD InStock
CA10037-156 RL200-508-M72
Anti-CD19 Rat Monoclonal Antibody (RPE (R-Phycoerythrin)) [clone: 1D3]
Antibodies
Anti-CD19 is useful for Flow Cytometry (Cell Surface). Researchers should determine optimal titers for applications that are not stated.

Recommended Dilutions: Flow Cytometry: 0.25-1µg/10^6 cells
Do not freeze! Light sensitive!

Type: Primary
Antigen: CD19
Clonality: Monoclonal
Clone: 1D3
Conjugation: RPE (R-Phycoerythrin)
Epitope:
Host: Rat
Isotype:
Reactivity: Mouse

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty