Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-IL2RA Rat Monoclonal Antibody (RPE (R-Phycoerythrin)) [clone: PC61]

Supplier: Rockland Immunochemicals
200-508-M73 200-508-M73
RL200-508-M73EA 405 USD InStock
RL200-508-M73 CA10037-158
Anti-IL2RA Rat Monoclonal Antibody (RPE (R-Phycoerythrin)) [clone: PC61]
Antibodies
Anti-CD25 is useful for Immunofluorescence and Flow Cytometry. Researchers should determine optimal titers for applications that are not stated.

Recommended Dilutions: Flow Cytometry: 0.2-1 µg/1x10^6 cells
Do not freeze! Light sensitive!

Type: Primary
Antigen: IL2RA
Clonality: Monoclonal
Clone: PC61
Conjugation: RPE (R-Phycoerythrin)
Epitope:
Host: Rat
Isotype: IgG1 lambda
Reactivity: Mouse

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty