Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-PTPRC Rat Monoclonal Antibody (RPE (R-Phycoerythrin)) [clone: 30-F11]

Supplier: Rockland Immunochemicals
200-508-N83 200-508-N83
RL200-508-N83EA 175 USD InStock
RL200-508-N83 CA200-508-N83
Anti-PTPRC Rat Monoclonal Antibody (RPE (R-Phycoerythrin)) [clone: 30-F11]
Antibodies
Anti-CD45 is useful for Flow Cytometry, Immunoprecipitation, immunochemistry, Immunofluorescence, and Western Blot using mouse spleen cells, or an appropriate cell type (where indicated).

Recommended Dilutions: Flow Cytometry: 10 µl/10^6 cells (0.1 µg);
Do not freeze! Light sensitive!

Type: Primary
Antigen: PTPRC
Clonality: Monoclonal
Clone: 30-F11
Conjugation: RPE (R-Phycoerythrin)
Epitope:
Host: Rat
Isotype: IgG2b kappa
Reactivity: Mouse

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty