Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-CD86 Rat Monoclonal Antibody (APC (Allophycocyanin)) [clone: GL-1]

Supplier: Rockland Immunochemicals
200-508-N90 200-508-N90
RL200-508-N90EA 405 USD InStock
CA200-508-N90 RL200-508-N90
Anti-CD86 Rat Monoclonal Antibody (APC (Allophycocyanin)) [clone: GL-1]
Antibodies
Anti-CD86 is useful for immunochemistry, Immunofluorescence, and Flow Cytometry using mouse spleen cells, or an appropriate cell type (where indicated). Researchers should determine optimal titers for applications that are not stated.

Recommended Dilutions: Flow Cytometry: 10 µl/10^6 cells (0.1 µg);
Do not freeze!

Type: Primary
Antigen: CD86
Clonality: Monoclonal
Clone: GL-1
Conjugation: APC (Allophycocyanin)
Epitope:
Host: Rat
Isotype: IgG2a kappa
Reactivity:

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty