Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-MDH1 Sheep Polyclonal Antibody (HRP (Horseradish Peroxidase))

Supplier: Rockland Immunochemicals
200-603-145S 200-603-145S
RL200-603-145SEA 148 USD InStock
RL200-603-145S CA10037-178
Anti-MDH1 Sheep Polyclonal Antibody (HRP (Horseradish Peroxidase))
Antibodies
Anti-Malate Dehydrogenase is useful in ELISA, western blot, and immunochemistry. Optimal titers should be obtained by researchers.

Recommended Dilutions: ELISA: 1:10000 - 1:50000; Western Blot: 1:1000 - 1:5000; Immunochemistry: 1:500 - 1:2500; Immunoprecipitation: 1:100;
Do not add Sodium Azide!

Type: Primary
Antigen: MDH1
Clonality: Polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Sheep
Isotype: IgG
Reactivity: Pig

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty