Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-ORF2 Chicken Polyclonal Antibody

Supplier: Rockland Immunochemicals
200-901-B20 200-901-B20S
RL200-901-B20EA 518 USD InStock
RL200-901-B20 RL200-901-B20S
Anti-ORF2 Chicken Polyclonal Antibody
Antibodies
This IgY fraction antibody has been tested for use in ELISA and western blot.  Specific conditions for reactivity should be optimized by the end user. Expect a band approximately 149 kDa in size corresponding to L1 Protein by western blotting.

Recommended Dilutions: ELISA: 1:10000; Western Blot: 1:2000 - 1:10000;

Type: Primary
Antigen: ORF2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Chicken
Isotype: IgG
Reactivity: Chimpanzee, Human

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty