Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-GFAP Chicken Polyclonal Antibody

Supplier: Rockland Immunochemicals
200-901-D60
RL200-901-D60EA 534 USD InStock
RL200-901-D60
Anti-GFAP Chicken Polyclonal Antibody
Antibodies
Anti-Glial Fibrillary Acidic Protein (Chicken) antibody is suitable for use in Western Blotting and IF. Specific conditions for reactivity should be optimized by the end user. Expect a band of approximately 50 kDa in size corresponding to the GFAP Protein.

Recommended Dilutions: Immunofluorescence: 1:1000; Western Blot: 1:10000

Type: Primary
Antigen: GFAP
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Chicken
Isotype: IgG
Reactivity: Bovine, Human, Mouse, Rat

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty