Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-MBP Chicken Polyclonal Antibody

Supplier: Rockland Immunochemicals
200-901-D81
RL200-901-D81EA 518 USD InStock
RL200-901-D81
Anti-MBP Chicken Polyclonal Antibody
Antibodies
Anti-Myelin Basic Protein antibody is suitable for use in Western Blotting and IF. Expect a bands of approximately 18 kDa and 22 kDa in size corresponding to MBP Proteins in Western blots of Rat brain homogenate.

Recommended Dilutions: Immunofluorescence: 1:500; Western Blot: 1:5000

Type: Primary
Antigen: MBP
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Chicken
Isotype: IgG
Reactivity: Bovine, Human, Mouse, Rat

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty