Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-HSPA1A Chicken Polyclonal Antibody

Supplier: Rockland Immunochemicals
200-901-F63
RL200-901-F63EA 501 USD InStock
RL200-901-F63
Anti-HSPA1A Chicken Polyclonal Antibody
Antibodies
Anti-Hsp70 Antibody is suitable for use in WB and ELISA. Specific conditions for reactivity should be optimized by the end user.

Recommended Dilutions: ELISA: 1:200; Western Blot: 1:1000

Type: Primary
Antigen: HSPA1A
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: Full length
Host: Chicken
Isotype: IgG
Reactivity: Human, Rat, Mouse

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty