Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-CD3E Armenian Hamster Monoclonal Antibody (FITC (Fluorescein Isothiocyanate)) [clone: 30-F11]

Supplier: Rockland Immunochemicals
200-B02-M69 200-B02-M69
RL200-B02-M69EA 161 USD InStock
RL200-B02-M69 CA10037-200
Anti-CD3E Armenian Hamster Monoclonal Antibody (FITC (Fluorescein Isothiocyanate)) [clone: 30-F11]
Antibodies
Anti-CD3 is useful for Flow Cytometry (Cell Surface). Researchers should determine optimal titers for applications that are not stated.

Recommended Dilutions: Flow Cytometry: 0.1-0.5 µg/10^6 cells; Sufficient to run approximately 500 tests
Do not freeze!

Type: Primary
Antigen: CD3E
Clonality: Monoclonal
Clone: 30-F11
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Armenian Hamster
Isotype: IgG
Reactivity:

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty