Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-TCRB Armenian Hamster Monoclonal Antibody (APC (Allophycocyanin)) [clone: H57-597]

Supplier: Rockland Immunochemicals
200-B26-N92 200-B26-N92
RL200-B26-N92EA 236 USD InStock
RL200-B26-N92 CA200-B26-N92
Anti-TCRB Armenian Hamster Monoclonal Antibody (APC (Allophycocyanin)) [clone: H57-597]
Antibodies
Anti-TCRbeta is useful for Immunoprecipitation, immunochemistry, and Flow Cytometry using mouse spleen cells, or an appropriate cell type (where indicated). Researchers should determine optimal titers for applications that are not stated.

Recommended Dilutions: Flow Cytometry: 10 µl/10^6 cells (0.1 µg);
Do not freeze!

Type: Primary
Antigen: TCRB
Clonality: Monoclonal
Clone: H57-597
Conjugation: APC (Allophycocyanin)
Epitope:
Host: Armenian Hamster
Isotype: IgG
Reactivity: Mouse

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty