Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-MSLN Mouse Monoclonal Antibody [clone: MN-1]

Supplier: Rockland Immunochemicals
200-301-A88S 200-301-A88S
RL200301A88SEA 148 USD InStock
RL200301A88S CA200301A88S
Anti-MSLN Mouse Monoclonal Antibody [clone: MN-1]
Antibodies
Type: Primary
Antigen: MSLN
Clonality: Monoclonal
Clone: MN-1
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty