Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-AKT3 Mouse Monoclonal Antibody [clone: 9A5-H9.G6]

Supplier: Rockland Immunochemicals
200-301-J36S 200-301-J36S
RL200301J36SEA 148 USD InStock
CA11029-198 RL200301J36S
Anti-AKT3 Mouse Monoclonal Antibody [clone: 9A5-H9.G6]
Antibodies
Type: Primary
Antigen: AKT3
Clonality: Monoclonal
Clone: 9A5-H9.G6
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty