Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-Human;Mouse;Rat;Monkey AKT Antibody [clone: 17F6.B11] (ATTO 594)

Supplier: Rockland Immunochemicals
200-355-268
RL200355268EA 508 USD InStock
RL200355268
Anti-Human;Mouse;Rat;Monkey AKT Antibody [clone: 17F6.B11] (ATTO 594)
Antibodies
Type: Secondary
Antigen: AKT
Clonality:
Clone: 17F6.B11
Conjugation: ATTO 594
Epitope:
Host:
Isotype:
Reactivity: Human, Mouse, Rat, Monkey

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty