Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-B2M Rabbit Polyclonal Antibody (Biotin)

Supplier: Rockland Immunochemicals
200-406-141-0100
RL200406141010EA 498 USD InStock
RL200406141010
Anti-B2M Rabbit Polyclonal Antibody (Biotin)
Antibodies
Type: Primary
Antigen: B2M
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty