Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-GOD Rabbit Polyclonal Antibody (Biotin)

Supplier: Rockland Immunochemicals
200-4637-0100 200-4637-0100
RL20046370100EA 498 USD InStock
RL20046370100 CA11029-284
Anti-GOD Rabbit Polyclonal Antibody (Biotin)
Antibodies
Type: Primary
Antigen: GOD
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype:
Reactivity: Aspergillus niger

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty