Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-CXCL10 Rabbit Polyclonal Antibody

Supplier: Rockland Immunochemicals
201-401-B84 201-401-B84S
RL201-401-B84SEA 148 USD InStock
RL201-401-B84S RL201-401-B84
Anti-CXCL10 Rabbit Polyclonal Antibody
Antibodies
This Protein-A purified antibody is suitable for ELISA and Western blotting. Specific conditions for reactivity should be optimized by the end user. Expect a band approximately 9.3 kDa in size corresponding to bovine CXCL10 Protein by western blotting.

Recommended Dilutions: ELISA: 1:10000; Western Blot: 1 µg/ml;

Type: Primary
Antigen: CXCL10
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity:

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty