Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-IgM µ Rabbit Polyclonal Antibody

Supplier: Rockland Immunochemicals
208-4107 208-4107
RL208-4107EA 594 USD InStock
CA208-4107 RL208-4107
Anti-IgM µ Rabbit Polyclonal Antibody
Antibodies
Secondary Rabbit Anti-IgM mu chain Reacts with Horse (Equine)

Recommended Dilutions: ELISA: 1:20000 - 1:100000; Western Blot: 1:2000 - 1:10000; Immunochemistry: 1:1000 - 1:5000;
Restore with deionized water (or equivalent); 5.0 ml

Type: Secondary
Antigen: IgM µ
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: µ-chain
Host: Rabbit
Isotype: IgM
Reactivity: Horse

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty