Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-THRB Mouse Monoclonal Antibody [clone: 2386]

Supplier: Rockland Immunochemicals
209-301-A96
RL209-301-A96EA 528 USD InStock
RL209-301-A96
Anti-THRB Mouse Monoclonal Antibody [clone: 2386]
Antibodies
Anti-Thyroid Hormone Receptor beta (Mouse) antibody is suitable for use in Western Blotting. Expect a band of approximately 55 kDa corresponding to TR--beta Proteins in the appropriate cell lysate or extract.

Recommended Dilutions: Western Blot: 1:1000

Type: Primary
Antigen: THRB
Clonality: Monoclonal
Clone: 2386
Conjugation: Unconjugated
Epitope: N-Terminal
Host: Mouse
Isotype: IgG1
Reactivity: Human

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty