Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-IL25 Mouse Monoclonal Antibody [clone: 11K8B1]

Supplier: Rockland Immunochemicals
209-301-C58 209-301-C58S
RL209-301-C58SEA 148 USD InStock
RL209-301-C58S RL209-301-C58
Anti-IL25 Mouse Monoclonal Antibody [clone: 11K8B1]
Antibodies
IL 17E antibody has been tested for use in Flow Cytometry and western blotting. Specific conditions for reactivity should be optimized by the end user.

Recommended Dilutions: ELISA: 1:10000-1:50000; Flow Cytometry: 1µg/ml; Western Blot: 0.5µg/ml;

Type: Primary
Antigen: IL25
Clonality: Monoclonal
Clone: 11K8B1
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG1
Reactivity: Human

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty