Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-cells in xenographic research Mouse Polyclonal Antibody [clone: 235-1]

Supplier: Rockland Immunochemicals
909-301-D99
RL209-301-D99EA 518 USD InStock
RL209-301-D99
Anti-cells in xenographic research Mouse Polyclonal Antibody [clone: 235-1]
Antibodies
Type: Primary
Antigen: cells in xenographic research
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty