Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-PGR Mouse Monoclonal Antibody [clone: 1154]

Supplier: Rockland Immunochemicals
209-301-E16
RL209-301-E16EA 528 USD InStock
RL209-301-E16
Anti-PGR Mouse Monoclonal Antibody [clone: 1154]
Antibodies
Anti-Progesterone Receptor pS190 (Mouse) antibody is suitable for use in Western Blot and IHC. Expect a band of approximately 90 kDa in size for PR-A isform and approximately 120 kDa in size for the PR-B isoform phosphorylated at Ser190 in Western Blotting.

Recommended Dilutions: Western Blot: 1:1000; Immunochemistry: 1:1000

Type: Primary
Antigen: PGR
Clonality: Monoclonal
Clone: 1154
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty