Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-PGR Mouse Monoclonal Antibody [clone: 608]

Supplier: Rockland Immunochemicals
209-301-E17
RL209-301-E17EA 528 USD InStock
RL209-301-E17
Anti-PGR Mouse Monoclonal Antibody [clone: 608]
Antibodies
Anti-Progesterone Receptor pS294 (Mouse) antibody is suitable for use in Western Blotting and IHC. Specific conditions for reactivity should but optimized by the end user. Expect a band of approximately 90 kDa in size.

Recommended Dilutions: Western Blot: 1:1000; Immunochemistry: 1:1000

Type: Primary
Antigen: PGR
Clonality: Monoclonal
Clone: 608
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty