Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-HSP90AA1 Mouse Monoclonal Antibody [clone: Hyb-K41009]

Supplier: Rockland Immunochemicals
209-301-F70
RL209-301-F70EA 532 USD InStock
RL209-301-F70
Anti-HSP90AA1 Mouse Monoclonal Antibody [clone: Hyb-K41009]
Antibodies
Anti-Hsp90 Antibody is suitable for WB, ELISA and IHC. Expect a band approximately 90kDa Proteins corresponding to the molecular mass of Hsp90a on SDS PAGE immunoblots. Specific conditions for reactivity should be optimized by the end user.

Recommended Dilutions: ELISA: 1:200; Western Blot: 1µg/ml

Type: Primary
Antigen: HSP90AA1
Clonality: Monoclonal
Clone: Hyb-K41009
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty