Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-HSP90AB1 Mouse Monoclonal Antibody [clone: Hyb-K3701]

Supplier: Rockland Immunochemicals
209-301-F71
RL209-301-F71EA 532 USD InStock
RL209-301-F71
Anti-HSP90AB1 Mouse Monoclonal Antibody [clone: Hyb-K3701]
Antibodies
Anti-Hsp90-beta Antibody is suitable for WB,ELISA, and IHC. Expect a band approximately 90kD Proteins corresponding to the molecular mass of hsp90-beta on SDS Page immunoblots. Specific conditions for reactivity should be optimized by the end user.

Recommended Dilutions: ELISA: 1:200; Western Blot: 1µg/ml

Type: Primary
Antigen: HSP90AB1
Clonality: Monoclonal
Clone: Hyb-K3701
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgM
Reactivity: Human

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty