Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-IL17F Mouse Monoclonal Antibody (Biotin) [clone: 4H1629.1]

Supplier: Rockland Immunochemicals
209-306-B31
RL209-306-B31EA 539 USD InStock
RL209-306-B31
Anti-IL17F Mouse Monoclonal Antibody (Biotin) [clone: 4H1629.1]
Antibodies
This purified antibody has been tested for use in Immunochemistryand Western Blot. Specific conditions for reactivity should be optimized by the end user.

Recommended Dilutions: ELISA: 1:10000-1:50000; Western Blot: 0.5 mg/ml; Immunochemistry: 5µg/ml;
Restore with deionized water (or equivalent); 100 µl

Type: Primary
Antigen: IL17F
Clonality: Monoclonal
Clone: 4H1629.1
Conjugation: Biotin
Epitope: Full length
Host: Mouse
Isotype: IgG1
Reactivity: Human

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty